Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOC3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022451.10 | NP_071896.8 | 2, 3 | 350 | ||
GenBank mRNA | AB077992.1 | - | 2, 3 | 237 | ||
AK091246.1 | - | 2, 3 | 307 | |||
AK292520.1 | - | 2, 3 | 332 | |||
BC049850.1 | - | 2, 3 | 300 | 5549517 | ||
BX505710.1 | - | 2, 3 | 352 |