Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ICA1 SIRNA Assays ›
Gene Symbol:
ICA1
Gene Name:
islet cell autoantigen 1
Gene Aliases:
ICA69, ICAp69
Chromosome Location:
Chr.7: 8113184 - 8262687 on Build GRCh38
UniGene ID:
Hs.487561
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
30066
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ICA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001136020.2
NP_001129492.1
5
579
NM_001276478.1
NP_001263407.1
5
561
NM_004968.3
NP_004959.2
5
564
NM_022307.2
NP_071682.1
5
379
XM_005249735.3
XP_005249792.1
5
501
XM_011515345.1
XP_011513647.1
5
579
XM_011515346.1
XP_011513648.1
5
564
XM_011515347.2
XP_011513649.1
5
451
XM_011515348.1
XP_011513650.1
5
497
XM_011515349.1
XP_011513651.1
5
430
XM_011515350.1
XP_011513652.1
5
501
XM_011515351.1
XP_011513653.1
5
636
XM_011515352.1
XP_011513654.1
5
455
XM_011515353.2
XP_011513655.1
5
508
XM_011515354.1
XP_011513656.1
2
92
XM_017012116.1
XP_016867605.1
5
347
XM_017012117.1
XP_016867606.1
5
537
XM_017012118.1
XP_016867607.1
5
497
XM_017012119.1
XP_016867608.1
5
472
XM_017012120.1
XP_016867609.1
5
518
XM_017012121.1
XP_016867610.1
5
385
XM_017012122.1
XP_016867611.1
6
764
XM_017012123.1
XP_016867612.1
6
703
XM_017012124.1
XP_016867613.1
6
864
XM_017012125.1
XP_016867614.1
6
614
GenBank mRNA
AK292106.1
-
5
448
BC008640.2
-
5
496
3854722
DB040266.1
-
5
507
EU416199.1
-
4
312
L01100.1
-
5
490
L21181.1
-
5
388
U26591.1
-
5
379
U26592.1
-
6
650
U37183.1
-
3
278
U38260.1
-
5
481
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
neurotransmitter transport
regulation of protein homodimerization activity
regulation of insulin secretion
molecular_function
protein binding
protein domain specific binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-7q954:80/10.239.162.42:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0