Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AZI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271650.1 | NP_001258579.1 | 9 | 1528 | ||
NM_022461.4 | NP_071906.1 | 8 | 1464 | |||
XM_005265392.2 | XP_005265449.1 | 9 | 1551 | |||
XM_011534009.2 | XP_011532311.1 | 8 | 1004 | |||
GenBank mRNA | AK025592.1 | - | 8 | 1311 | ||
AK290637.1 | - | 8 | 1183 | |||
AY151386.1 | - | 7 | 932 | |||
BM546093.1 | - | 7 | 800 | |||
BX648471.1 | - | 9 | 1192 |