Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CLEC7A SIRNA Assays ›
Gene Symbol:
CLEC7A
Gene Name:
C-type lectin domain family 7 member A
Gene Aliases:
BGR, CANDF4, CD369, CLECSF12, DECTIN1, SCARE2
Chromosome Location:
Chr.12: 10116777 - 10130269 on Build GRCh38
UniGene ID:
Hs.143929
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
30145
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CLEC7A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022570.4
NP_072092.2
1
275
NM_197947.2
NP_922938.1
1
275
NM_197948.2
NP_922939.1
1
275
NM_197949.2
NP_922940.1
1
275
NM_197950.2
NP_922941.1
1
275
NM_197954.2
NP_922945.1
1
275
NR_125336.1
1
275
XM_006719135.3
XP_006719198.1
1
270
XM_017019822.1
XP_016875311.1
1
269
XM_017019823.1
XP_016875312.1
1
172
GenBank mRNA
AA361063.1
-
1
110
AF313468.1
-
2
139
AF313469.1
-
1
138
AF400595.1
-
1
88
AF400596.1
-
1
88
AF400597.1
-
1
88
AF400599.1
-
1
88
AF400600.1
-
1
88
AF400601.1
-
1
88
AF400602.1
-
1
88
AJ312372.1
-
1
243
AJ312373.1
-
1
243
AK294111.1
-
1
267
AK295108.1
-
1
219
AK297028.1
-
1
159
AK298679.1
-
1
191
AK298724.1
-
1
191
AK313247.1
-
1
206
AY009090.2
-
1
159
AY026769.2
-
1
88
AY026770.2
-
1
88
AY026771.1
-
1
88
AY359002.1
-
1
247
BC013385.1
-
1
144
BC071746.1
-
1
189
4690475
BC093829.1
-
1
164
BC093831.1
-
1
164
CR749536.1
-
1
218
DA193873.1
-
1
275
KU178745.1
-
1
88
KU178746.1
-
1
88
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
pattern recognition receptor signaling pathway
stimulatory C-type lectin receptor signaling pathway
leukocyte activation involved in immune response
phagocytosis, recognition
inflammatory response
cell recognition
carbohydrate mediated signaling
T cell activation
defense response to protozoan
innate immune response
cellular response to molecule of fungal origin
protein binding
signaling pattern recognition receptor activity
carbohydrate binding
MHC protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-462rt:80/10.239.167.23:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0