Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022733.2 | NP_073570.1 | 1 | 489 | ||
XM_011541960.1 | XP_011540262.1 | 1 | 433 | |||
XM_017002093.1 | XP_016857582.1 | 1 | 487 | |||
GenBank mRNA | AK313105.1 | - | 1 | 436 | ||
AL137764.1 | - | 1 | 16 | |||
BC021133.1 | - | 1 | 139 | 4855320 |