Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EWSR1 SIRNA Assays ›
Gene Symbol:
EWSR1
Gene Name:
EWS RNA binding protein 1
Gene Aliases:
EWS, EWS-FLI1, bK984G1.4
Chromosome Location:
Chr.22: 29268009 - 29300525 on Build GRCh38
UniGene ID:
Hs.374477
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
3017
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EWSR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001163285.1
NP_001156757.1
13
1642
NM_001163286.1
NP_001156758.1
12
1477
NM_005243.3
NP_005234.1
13
1645
NM_013986.3
NP_053733.2
14
1660
XM_005261389.3
XP_005261446.1
13
1400
XM_005261390.4
XP_005261447.1
12
1232
XM_011529995.2
XP_011528297.1
13
1375
XM_011529996.2
XP_011528298.1
13
1372
XM_011529997.1
XP_011528299.1
13
1375
XM_011529998.1
XP_011528300.1
13
1372
XM_011529999.2
XP_011528301.1
13
1375
XM_011530000.1
XP_011528302.1
13
1375
XM_011530001.2
XP_011528303.1
12
1207
XM_011530002.2
XP_011528304.1
11
1156
XM_017028644.1
XP_016884133.1
13
1397
XM_017028645.1
XP_016884134.1
13
1400
XM_017028646.1
XP_016884135.1
13
1372
XM_017028647.1
XP_016884136.1
13
1397
XM_017028648.1
XP_016884137.1
13
1397
XM_017028654.1
XP_016884143.1
12
1204
XM_017028655.1
XP_016884144.1
12
1229
XM_017028657.1
XP_016884146.1
13
5126
XM_017028658.1
XP_016884147.1
12
1204
XM_017028659.1
XP_016884148.1
12
1229
XM_017028661.1
XP_016884150.1
13
5123
XM_017028662.1
XP_016884151.1
12
1229
XM_017028664.1
XP_016884153.1
12
1317
XM_017028665.1
XP_016884154.1
12
1317
XM_017028666.1
XP_016884155.1
12
5088
GenBank mRNA
AK026270.1
-
4
1613
AK056309.1
-
14
1353
AK056681.1
-
12
1160
AK096787.1
-
2
866
AK127624.1
-
9
1198
AK225096.1
-
13
1324
AK225102.1
-
13
1336
AL833489.1
-
5
3269
BC004817.1
-
13
1336
3449145
BC011048.1
-
13
1341
4340116
BC072442.1
-
12
1320
6047805
BE741903.1
-
4
342
BX648769.1
-
8
6267
CR456490.1
-
13
1347
JX977847.1
-
13
1537
X66899.1
-
13
1360
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
nucleotide binding
RNA binding
protein binding
calmodulin binding
zinc ion binding
identical protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0