Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022831.2 | NP_073742.2 | 1 | 303 | ||
GenBank mRNA | AK022868.1 | - | 1 | 255 | ||
AK289865.1 | - | 1 | 254 | |||
AL833718.1 | - | 1 | 93 | |||
BC015535.1 | - | 1 | 230 | 3903579 | ||
BC043142.1 | - | 1 | 522 | |||
BC067805.1 | - | 1 | 251 | 5298783 | ||
BC171848.1 | - | 1 | 278 | |||
DA483113.1 | - | 1 | 252 |