Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TNS1 SIRNA Assays ›
Gene Symbol:
TNS1
Gene Name:
tensin 1
Gene Aliases:
MST091, MST122, MST127, MSTP091, MSTP122, MSTP127, MXRA6, PPP1R155, TNS
Chromosome Location:
Chr.2: 217799789 - 217978853 on Build GRCh38
UniGene ID:
Hs.471381
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
30337
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TNS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308022.1
NP_001294951.1
31
6946
NM_001308023.1
NP_001294952.1
30
6925
NM_022648.5
NP_072174.3
33
7197
XM_011511712.1
XP_011510014.1
31
7180
XM_011511713.1
XP_011510015.1
31
7165
XM_011511715.1
XP_011510017.1
30
7141
XM_011511723.2
XP_011510025.1
31
7141
XM_011511728.1
XP_011510030.1
15
8233
XM_017004801.1
XP_016860290.1
32
7333
XM_017004802.1
XP_016860291.1
31
7288
XM_017004803.1
XP_016860292.1
31
7249
XM_017004804.1
XP_016860293.1
32
7204
XM_017004805.1
XP_016860294.1
31
7120
XM_017004806.1
XP_016860295.1
30
7093
XM_017004807.1
XP_016860296.1
29
7054
XM_017004808.1
XP_016860297.1
28
6829
XM_017004809.1
XP_016860298.1
36
7677
XM_017004810.1
XP_016860299.1
36
7560
XM_017004811.1
XP_016860300.1
34
7423
XM_017004812.1
XP_016860301.1
33
7373
XM_017004813.1
XP_016860302.1
19
6285
XM_017004814.1
XP_016860303.1
19
6073
XM_017004815.1
XP_016860304.1
19
6139
XM_017004816.1
XP_016860305.1
12
6448
XM_017004817.1
XP_016860306.1
17
8428
GenBank mRNA
AB209238.1
-
15
4213
AF116610.1
-
1
1418
AL117537.1
-
9
2405
BC014042.2
-
10
2909
Back To Top
More Information
Panther Classification:
Molecular Function -
non-motor actin binding protein
Gene Ontology Categories:
Function(s)
Process(es)
cell-substrate junction assembly
fibroblast migration
actin binding
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0