Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198978.1 | NP_001185907.1 | 10 | 2166 | ||
NM_001198979.1 | NP_001185908.1 | 10 | 2250 | |||
NM_001198980.1 | NP_001185909.1 | 10 | 2181 | |||
NM_022733.2 | NP_073570.1 | 10 | 2608 | |||
XM_011541960.1 | XP_011540262.1 | 11 | 2615 | |||
XM_017002093.1 | XP_016857582.1 | 7 | 2307 | |||
GenBank mRNA | AL133206.1 | - | 10 | 2122 | ||
AL137764.1 | - | 10 | 2135 | |||
BC014512.1 | - | 8 | 1947 | |||
BC021133.1 | - | 10 | 2258 | 4855320 |