Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRTC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042574.2 | NP_001036039.1 | 14 | 1778 | ||
NM_022769.4 | NP_073606.3 | 14 | 1778 | |||
XM_005254968.2 | XP_005255025.1 | 14 | 1746 | |||
XM_011521906.2 | XP_011520208.1 | 13 | 1650 | |||
GenBank mRNA | AK025521.1 | - | 7 | 945 | ||
AK074170.1 | - | 11 | 1249 | |||
AK090443.1 | - | 14 | 1738 | |||
AK225300.1 | - | 7 | 945 | |||
AY360173.1 | - | 14 | 1599 | |||
BC074730.2 | - | 5 | 721 | |||
BC074731.2 | - | 5 | 721 | 30915456 | ||
BP365179.1 | - | 4 | 523 | |||
CN298837.1 | - | 2 | 93 |