Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPS8L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022772.3 | NP_073609.2 | 9, 10 | 1017 | ||
XM_017018130.1 | XP_016873619.1 | 10, 11 | 1234 | |||
XM_017018131.1 | XP_016873620.1 | 9, 10 | 1105 | |||
XM_017018132.1 | XP_016873621.1 | 10, 11 | 1075 | |||
GenBank mRNA | AK025824.1 | - | 9, 10 | 935 | ||
AK122903.1 | - | 6, 7 | 1213 | |||
AK222903.1 | - | 9, 10 | 930 | |||
BC101481.1 | - | 8, 9 | 810 | 8068987 |