- Order Status
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: AIDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022831.2 | NP_073742.2 | 10 | 2164 | ||
GenBank mRNA | AF131838.1 | - | 1 | 604 | ||
AK022868.1 | - | 10 | 2116 | |||
AL833718.1 | - | 10 | 1954 | |||
BC043142.1 | - | 10 | 3212 |