Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM129B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035534.2 | NP_001030611.1 | 13 | 1625 | ||
NM_022833.3 | NP_073744.2 | 13 | 1837 | |||
XM_005252135.2 | XP_005252192.2 | 14 | 1684 | |||
XM_011518925.1 | XP_011517227.1 | 14 | 1856 | |||
GenBank mRNA | AB210016.1 | - | 13 | 1575 | ||
AF151783.1 | - | 13 | 1607 | |||
AF192911.1 | - | 13 | 1828 | |||
AK023580.1 | - | 5 | 636 | |||
AL137555.1 | - | 12 | 1445 | |||
BC001979.1 | - | 6 | 724 | |||
BC110345.1 | - | 4 | 337 |