Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCLRE1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022836.3 | NP_073747.1 | 2 | 630 | ||
GenBank mRNA | AF306693.1 | - | 2 | 565 | ||
AK022872.1 | - | 2 | 176 | |||
AK024060.1 | - | 2 | 548 | |||
BC029687.1 | - | 2 | 507 | 5001181 |