Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RANBP17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022897.4 | NP_075048.1 | 2 | 247 | ||
XM_011534627.1 | XP_011532929.1 | 2 | 103 | |||
XM_011534631.2 | XP_011532933.1 | 1 | 116 | |||
XM_011534636.2 | XP_011532938.1 | 2 | 103 | |||
XM_011534637.2 | XP_011532939.1 | 2 | 103 | |||
XM_017009736.1 | XP_016865225.1 | 2 | 103 | |||
XM_017009737.1 | XP_016865226.1 | 1 | 116 | |||
XM_017009738.1 | XP_016865227.1 | 2 | 103 | |||
XM_017009739.1 | XP_016865228.1 | 2 | 103 | |||
XM_017009740.1 | XP_016865229.1 | 2 | 103 | |||
XM_017009741.1 | XP_016865230.1 | 2 | 103 | |||
XM_017009742.1 | XP_016865231.1 | 2 | 103 | |||
XM_017009743.1 | XP_016865232.1 | 2 | 277 | |||
XM_017009744.1 | XP_016865233.1 | 2 | 103 | |||
XM_017009746.1 | XP_016865235.1 | 2 | 103 | |||
GenBank mRNA | AF222747.1 | - | 2 | 103 | ||
AJ271459.1 | - | 2 | 111 | |||
AJ288952.1 | - | 2 | 111 | |||
AJ288953.1 | - | 2 | 111 | |||
AJ288954.1 | - | 2 | 111 | |||
AJ288955.1 | - | 2 | 111 | |||
AK298785.1 | - | 2 | 260 | |||
AK315159.1 | - | 2 | 197 | |||
AL832980.1 | - | 2 | 94 | |||
DC365649.1 | - | 2 | 260 | |||
DC404010.1 | - | 2 | 247 |