Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8orf33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023080.2 | NP_075568.1 | 3 | 398 | ||
GenBank mRNA | AK024642.1 | - | 3 | 396 | ||
BC010001.2 | - | 3 | 358 | 4135341 | ||
BC015181.1 | - | 2 | 159 | |||
BX648996.1 | - | 2 | 458 | |||
CR457330.1 | - | 2 | 344 | |||
DA467518.1 | - | 3 | 398 |