Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRVI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098579.2 | NP_001092049.2 | 11, 12 | 1656 | ||
NM_001100163.2 | NP_001093633.1 | 12, 13 | 1934 | |||
NM_001100167.2 | NP_001093637.1 | 12, 13 | 1651 | |||
NM_001206880.1 | NP_001193809.1 | 11, 12 | 1369 | |||
NM_001206881.1 | NP_001193810.1 | 10, 11 | 1776 | |||
NM_130385.3 | NP_569056.4 | 12, 13 | 1987 | |||
GenBank mRNA | AF081249.1 | - | 10, 11 | 1672 | ||
AF081250.1 | - | 10, 11 | 1400 | |||
AK127209.1 | - | 12, 13 | 1629 | |||
AK296336.1 | - | 12, 13 | 1751 | |||
AK300358.1 | - | 11, 12 | 1641 | |||
AK301296.1 | - | 11, 12 | 980 | |||
AK304702.1 | - | 11, 12 | 1003 | |||
BC117127.1 | - | 12, 13 | 1524 | 40125678 |