Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM6SF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144903.1 | NP_001138375.1 | 4, 5 | 516 | ||
NM_023003.3 | NP_075379.2 | 4, 5 | 516 | |||
XM_011521679.2 | XP_011519981.1 | 4, 5 | 441 | |||
XM_011521680.2 | XP_011519982.1 | 4, 5 | 441 | |||
XM_017022320.1 | XP_016877809.1 | 4, 5 | 441 | |||
XM_017022321.1 | XP_016877810.1 | 4, 5 | 441 | |||
XM_017022322.1 | XP_016877811.1 | 4, 5 | 441 | |||
XM_017022323.1 | XP_016877812.1 | 4, 5 | 568 | |||
XM_017022324.1 | XP_016877813.1 | 4, 5 | 545 | |||
XM_017022328.1 | XP_016877817.1 | 4, 5 | 511 | |||
XM_017022329.1 | XP_016877818.1 | 4, 5 | 501 | |||
XM_017022330.1 | XP_016877819.1 | 4, 5 | 501 | |||
XM_017022331.1 | XP_016877820.1 | 4, 5 | 501 | |||
GenBank mRNA | AF255922.1 | - | 4, 5 | 464 | ||
AK292100.1 | - | 4, 5 | 441 | |||
AK295568.1 | - | 4, 5 | 405 | |||
BC063487.1 | - | 4, 5 | 516 | 5240735 | ||
DQ021909.1 | - | 4, 5 | 407 |