Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGLEC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023068.3 | NP_075556.1 | 16 | 4263 | ||
XM_006723610.3 | XP_006723673.1 | 17 | 4458 | |||
XM_011529324.2 | XP_011527626.1 | 17 | 4455 | |||
XM_011529325.2 | XP_011527627.1 | 17 | 4461 | |||
XM_011529326.2 | XP_011527628.1 | 17 | 4457 | |||
XM_011529327.2 | XP_011527629.1 | 16 | 4391 | |||
XM_011529328.2 | XP_011527630.1 | 17 | 4403 | |||
XM_011529329.1 | XP_011527631.1 | 16 | 5644 | |||
XM_011529330.1 | XP_011527632.1 | 15 | 5386 | |||
XM_011529331.1 | XP_011527633.1 | 16 | 5562 | |||
XM_011529332.1 | XP_011527634.1 | 15 | 3848 | |||
GenBank mRNA | AF230073.1 | - | 16 | 4263 | ||
AK024459.1 | - | 3 | 703 | |||
AK024462.1 | - | 8 | 2067 | |||
AK024479.1 | - | 3 | 703 | |||
AK122586.1 | - | 5 | 4234 | |||
BC141884.1 | - | 16 | 4263 |