Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024057.3 | NP_076962.2 | 5, 6 | 604 | ||
GenBank mRNA | AF514994.1 | - | 4, 5 | 458 | ||
AK026271.1 | - | 4, 5 | 439 | |||
AK296733.1 | - | 5, 6 | 562 | |||
AK310619.1 | - | 5, 6 | 564 | |||
BC000861.2 | - | 4, 5 | 522 | 3459639 |