Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMIM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024104.3 | NP_077009.2 | 5 | 609 | ||
GenBank mRNA | AK025602.1 | - | 5 | 566 | ||
BC001680.2 | - | 2 | 381 | |||
BC001948.1 | - | 2 | 381 | |||
BC069279.1 | - | 6 | 939 | |||
BX648276.1 | - | 4 | 506 |