Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024333.2 | NP_077309.1 | 3 | 351 | ||
XM_005259644.2 | XP_005259701.1 | 3 | 350 | |||
GenBank mRNA | AF316829.1 | - | 3 | 242 | ||
AK021750.1 | - | 3 | 251 | |||
AK295523.1 | - | 3 | 246 | |||
AK303606.1 | - | 3 | 235 | |||
AK315661.1 | - | 3 | 345 | |||
AY032617.1 | - | 3 | 274 | |||
BC003124.1 | - | 3 | 251 | 3502614 |