Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ODF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024410.3 | NP_077721.2 | 1 | 391 | ||
GenBank mRNA | AK315276.1 | - | 1 | 384 | ||
BC104456.1 | - | 1 | 362 | 40030623 | ||
BC104457.1 | - | 1 | 362 | 40030627 | ||
DB457574.1 | - | 1 | 391 |