Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSEN34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077446.3 | NP_001070914.1 | 4 | 1368 | ||
NM_001282332.1 | NP_001269261.1 | 5 | 1528 | |||
NM_001282333.1 | NP_001269262.1 | 6 | 1022 | |||
NM_024075.4 | NP_076980.2 | 5 | 1404 | |||
XM_005278290.4 | XP_005278347.1 | 5 | 1320 | |||
XM_011527294.2 | XP_011525596.1 | 5 | 1299 | |||
XM_011527295.1 | XP_011525597.1 | 5 | 1301 | |||
GenBank mRNA | AF211970.1 | - | 2 | 529 | ||
AK025929.1 | - | 5 | 1323 | |||
AK054944.1 | - | 3 | 2354 | |||
AK225385.1 | - | 5 | 1323 | |||
AK226013.1 | - | 5 | 1326 | |||
AK300054.1 | - | 6 | 1003 | |||
BC004530.2 | - | 5 | 1528 | 3342968 | ||
BC020805.1 | - | 4 | 1308 | 4277870 |