Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024527.4 | NP_078803.4 | 2 | 824 | ||
GenBank mRNA | AK021805.1 | - | 2 | 776 | ||
AK225140.1 | - | 2 | 750 | |||
AK225340.1 | - | 2 | 777 | |||
AK298251.1 | - | 3 | 706 | |||
AK315366.1 | - | 2 | 764 | |||
BC007895.2 | - | 2 | 726 | 4131888 | ||
BC020173.1 | - | 2 | 626 | 3542724 | ||
BC039087.1 | - | 2 | 669 | 5165152 | ||
EU285559.1 | - | 1 | 584 |