Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCRN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193528.1 | NP_001180457.1 | 6 | 834 | ||
NM_024583.4 | NP_078859.2 | 6 | 925 | |||
XM_005246853.2 | XP_005246910.1 | 6 | 1330 | |||
XM_005246854.4 | XP_005246911.1 | 6 | 2490 | |||
XM_005246855.2 | XP_005246912.1 | 6 | 1378 | |||
XM_005246856.2 | XP_005246913.1 | 6 | 1334 | |||
XM_005246857.2 | XP_005246914.1 | 6 | 1034 | |||
XM_011511839.2 | XP_011510141.1 | 5 | 734 | |||
XM_017004909.1 | XP_016860398.1 | 6 | 1922 | |||
XM_017004910.1 | XP_016860399.1 | 6 | 1400 | |||
GenBank mRNA | AF279776.1 | - | 2 | 262 | ||
AK026795.1 | - | 2 | 272 | |||
AK057912.1 | - | 6 | 903 | |||
AK295357.1 | - | 6 | 818 | |||
AK296558.1 | - | 5 | 724 | |||
BC031821.1 | - | 3 | 386 | |||
BC119684.1 | - | 6 | 876 | 40116568 | ||
BC119685.1 | - | 6 | 876 | 40116570 |