Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SAP130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145928.1 | NP_001139400.1 | 21 | 3923 | ||
NM_024545.3 | NP_078821.2 | 20 | 3818 | |||
XM_005263767.3 | XP_005263824.1 | 21 | 3894 | |||
XM_005263768.3 | XP_005263825.1 | 21 | 3873 | |||
XM_005263769.3 | XP_005263826.1 | 21 | 3794 | |||
XM_005263770.3 | XP_005263827.1 | 21 | 3870 | |||
XM_006712748.3 | XP_006712811.1 | 21 | 4080 | |||
XM_006712749.3 | XP_006712812.1 | 20 | 3975 | |||
XM_017004903.1 | XP_016860392.1 | 21 | 3870 | |||
XM_017004904.1 | XP_016860393.1 | 20 | 3768 | |||
XM_017004905.1 | XP_016860394.1 | 20 | 3765 | |||
GenBank mRNA | AK022823.1 | - | 11 | 2440 | ||
AK092234.1 | - | 14 | 2876 | |||
AL136833.1 | - | 20 | 3816 | |||
BC017453.1 | - | 13 | 2623 | |||
DB509348.1 | - | 1 | 134 |