Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C14orf169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024644.4 | NP_078920.2 | 1 | 196 | ||
GenBank mRNA | AB101204.1 | - | 1 | 303 | ||
AK025455.1 | - | 1 | 166 | |||
AK299994.1 | - | 1 | 176 | |||
AY390535.1 | - | 1 | 163 | |||
BC011350.1 | - | 1 | 165 | 4340013 | ||
BC071954.1 | - | 1 | 143 | 6728391 | ||
DA509240.1 | - | 1 | 176 | |||
DA872764.1 | - | 1 | 195 |