Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278647.1 | NP_001265576.1 | 2 | 250 | ||
NM_001278648.1 | NP_001265577.1 | 2 | 271 | |||
NM_005341.3 | NP_005332.1 | 2 | 337 | |||
XM_017001110.1 | XP_016856599.1 | 2 | 322 | |||
XM_017001111.1 | XP_016856600.1 | 2 | 322 | |||
XM_017001112.1 | XP_016856601.1 | 2 | 327 | |||
XM_017001113.1 | XP_016856602.1 | 2 | 268 | |||
XM_017001114.1 | XP_016856603.1 | 2 | 255 | |||
GenBank mRNA | AK290156.1 | - | 2 | 245 | ||
AK312786.1 | - | 2 | 253 | |||
BC013573.2 | - | 1 | 196 | 3906038 | ||
BI913659.1 | - | 2 | 312 | |||
BP281477.1 | - | 2 | 260 | |||
HY047768.1 | - | 2 | 250 | |||
L16896.1 | - | 2 | 277 |