Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VEPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167911.1 | NP_001161383.1 | 13 | 2757 | ||
NM_001167912.1 | NP_001161384.1 | 14 | 2892 | |||
NM_024621.2 | NP_078897.2 | 14 | 3083 | |||
XM_011513134.2 | XP_011511436.1 | 13 | 2826 | |||
XM_011513135.1 | XP_011511437.1 | 14 | 2722 | |||
GenBank mRNA | AB051479.1 | - | 14 | 2892 | ||
AK022666.1 | - | 4 | 760 | |||
AK027625.1 | - | 14 | 2981 | |||
AL713656.1 | - | 13 | 2681 | |||
BC026235.1 | - | 6 | 1205 | |||
BC042159.1 | - | 7 | 1542 | |||
BC111017.1 | - | 13 | 2750 | 5174889 |