Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C14orf169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024644.4 | NP_078920.2 | 1 | 2212 | ||
GenBank mRNA | AB101204.1 | - | 1 | 2322 | ||
AK025455.1 | - | 1 | 2184 | |||
AK299994.1 | - | 2 | 1878 | |||
AY390535.1 | - | 1 | 2182 | |||
BC011350.1 | - | 1 | 2184 | 4340013 | ||
BC071954.1 | - | 1 | 2160 | 6728391 |
Gene Symbol: ACOT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017021591.1 | XP_016877080.1 | 1 | 125 |