Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GTDC1 SIRNA Assays ›
Gene Symbol:
GTDC1
Gene Name:
glycosyltransferase like domain containing 1
Gene Aliases:
Hmat-Xa, mat-Xa
Chromosome Location:
Chr.2: 143945787 - 144333092 on Build GRCh38
UniGene ID:
Hs.44780
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
31463
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GTDC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001006636.3
NP_001006637.1
12
2312
NM_001164629.3
NP_001158101.1
12
2297
NM_001284233.1
NP_001271162.1
9
1976
NM_001284234.1
NP_001271163.1
10
2012
NM_001284238.1
NP_001271167.1
14
2511
NM_024659.4
NP_078935.2
10
2024
XM_005263774.2
XP_005263831.1
13
2396
XM_005263782.2
XP_005263839.1
13
2412
XM_005263784.2
XP_005263841.1
11
2271
XM_011511843.2
XP_011510145.1
13
2445
XM_011511855.2
XP_011510157.1
11
2389
XM_011511856.2
XP_011510158.1
11
2171
XM_017004917.1
XP_016860406.1
17
3067
XM_017004918.1
XP_016860407.1
15
2659
XM_017004919.1
XP_016860408.1
13
2545
XM_017004920.1
XP_016860409.1
15
5701
XM_017004921.1
XP_016860410.1
14
2494
XM_017004922.1
XP_016860411.1
14
2671
XM_017004923.1
XP_016860412.1
13
2459
XM_017004924.1
XP_016860413.1
15
2655
XM_017004925.1
XP_016860414.1
12
2317
XM_017004926.1
XP_016860415.1
10
2248
XM_017004927.1
XP_016860416.1
12
2279
XM_017004928.1
XP_016860417.1
14
2512
XM_017004929.1
XP_016860418.1
12
2249
XM_017004930.1
XP_016860419.1
12
2900
XM_017004931.1
XP_016860420.1
12
2313
XM_017004932.1
XP_016860421.1
12
2283
XM_017004933.1
XP_016860422.1
12
2265
XM_017004934.1
XP_016860423.1
13
5472
XM_017004935.1
XP_016860424.1
14
3342
XM_017004936.1
XP_016860425.1
13
2519
XM_017004937.1
XP_016860426.1
10
2255
XM_017004938.1
XP_016860427.1
9
2143
XM_017004939.1
XP_016860428.1
12
2303
XM_017004940.1
XP_016860429.1
9
2104
XM_017004941.1
XP_016860430.1
8
1993
XM_017004942.1
XP_016860431.1
11
2150
XM_017004943.1
XP_016860432.1
11
2816
XM_017004944.1
XP_016860433.1
10
2039
XM_017004945.1
XP_016860434.1
12
2257
XM_017004946.1
XP_016860435.1
10
1994
XM_017004947.1
XP_016860436.1
10
2677
XM_017004948.1
XP_016860437.1
11
5217
XM_017004949.1
XP_016860438.1
9
1893
XM_017004950.1
XP_016860439.1
11
1940
XM_017004951.1
XP_016860440.1
11
1895
XM_017004952.1
XP_016860441.1
11
2339
XM_017004953.1
XP_016860442.1
11
1911
XM_017004954.1
XP_016860443.1
12
1999
XM_017004955.1
XP_016860444.1
14
2347
XM_017004956.1
XP_016860445.1
8
1750
XM_017004957.1
XP_016860446.1
10
1796
GenBank mRNA
AB112440.1
-
1
69
AK021815.1
-
6
1488
AK091684.1
-
9
1980
AY281366.1
-
12
2312
BC017741.2
-
11
2064
4390876
BX647773.1
-
14
2450
Back To Top
More Information
Panther Classification:
Molecular Function -
glycosyltransferase
transferase
metabolite interconversion enzyme
Gene Ontology Categories:
Process(es)
transferase activity, transferring glycosyl groups
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-k9xzd:80/10.239.163.240:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0