Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024712.3 | NP_078988.2 | 15 | 1686 | ||
GenBank mRNA | AK023886.1 | - | 14 | 1450 | ||
AK056923.1 | - | 14 | 1649 | |||
AK300976.1 | - | 15 | 1700 | |||
BC015524.1 | - | 5 | 496 | |||
BC034410.1 | - | 12 | 1298 |