Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KATNBL1P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003954.1 | 1 | 982 | |||
GenBank mRNA | BC067801.1 | - | 1 | 982 |
Gene Symbol: KATNBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024713.2 | NP_078989.1 | 9 | 1027 | ||
XM_005254668.3 | XP_005254725.1 | 9 | 1077 | |||
XM_017022571.1 | XP_016878060.1 | 10 | 1111 | |||
XM_017022572.1 | XP_016878061.1 | 10 | 1416 | |||
XM_017022573.1 | XP_016878062.1 | 9 | 1237 | |||
XM_017022574.1 | XP_016878063.1 | 10 | 1090 | |||
XM_017022575.1 | XP_016878064.1 | 6 | 799 | |||
GenBank mRNA | AK026210.1 | - | 9 | 846 | ||
AK293021.1 | - | 9 | 964 | |||
AL136908.1 | - | 9 | 953 | |||
AL833631.1 | - | 9 | 1011 | |||
BC026234.1 | - | 9 | 919 | 4693115 | ||
BC111000.1 | - | 9 | 988 | 5751934 | ||
BX389537.2 | - | 3 | 204 |