Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RERGL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286201.1 | NP_001273130.1 | 2 | 126 | ||
NM_024730.3 | NP_079006.1 | 3 | 283 | |||
NR_104413.1 | 2 | 126 | ||||
GenBank mRNA | AA220227.1 | - | 2 | 74 | ||
AK026308.1 | - | 3 | 209 | |||
BC042888.1 | - | 3 | 270 | 5198817 | ||
BX422875.2 | - | 2 | 74 | |||
DT220095.1 | - | 3 | 283 |