Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105248.1 | NP_001098718.1 | 14, 15 | 3021 | ||
NM_001105249.1 | NP_001098719.1 | 14, 15 | 3021 | |||
NM_001261841.1 | NP_001248770.1 | 14, 15 | 2976 | |||
NM_001308161.1 | NP_001295090.1 | 13, 14 | 2658 | |||
NM_024780.4 | NP_079056.2 | 10, 11 | 1791 | |||
GenBank mRNA | AK023655.1 | - | 3, 4 | 274 | ||
AK093944.1 | - | 7, 8 | 848 | |||
AK097718.1 | - | 10, 11 | 1756 | |||
AK131202.1 | - | 11, 12 | 1511 | |||
AK299055.1 | - | 13, 14 | 2658 | |||
AK304431.1 | - | 14, 15 | 2976 | |||
AY236494.1 | - | 10, 11 | 1791 | |||
AY263164.1 | - | 10, 11 | 1534 | |||
AY358155.1 | - | 12, 13 | 2370 | |||
BC027602.1 | - | 10, 11 | 1746 | 4839111 | ||
BC038118.2 | - | 10, 11 | 1486 | 5265527 | ||
CR749359.1 | - | 12, 13 | 1676 |