Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105248.1 | NP_001098718.1 | 15 | 3064 | ||
NM_001105249.1 | NP_001098719.1 | 15 | 3064 | |||
NM_001261841.1 | NP_001248770.1 | 15 | 3019 | |||
NM_001308161.1 | NP_001295090.1 | 14 | 2701 | |||
NM_024780.4 | NP_079056.2 | 11 | 1834 | |||
GenBank mRNA | AK023655.1 | - | 4 | 317 | ||
AK093944.1 | - | 8 | 891 | |||
AK097718.1 | - | 11 | 1799 | |||
AK131202.1 | - | 12 | 1554 | |||
AK299055.1 | - | 14 | 2701 | |||
AK304431.1 | - | 15 | 3019 | |||
AY236494.1 | - | 11 | 1834 | |||
AY263164.1 | - | 11 | 1577 | |||
AY358155.1 | - | 13 | 2413 | |||
BC027602.1 | - | 11 | 1789 | 4839111 | ||
BC038118.2 | - | 11 | 1529 | 5265527 | ||
CR749359.1 | - | 13 | 1719 |