Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD13A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024789.3 | NP_079065.2 | 10 | 2303 | ||
XM_006717973.3 | XP_006718036.1 | 12 | 2506 | |||
XM_006717974.3 | XP_006718037.1 | 9 | 2242 | |||
XM_011540160.2 | XP_011538462.1 | 11 | 2926 | |||
XM_011540163.2 | XP_011538465.1 | 11 | 2416 | |||
XM_011540167.2 | XP_011538469.1 | 8 | 1729 | |||
XM_017016654.1 | XP_016872143.1 | 11 | 2460 | |||
XM_017016655.1 | XP_016872144.1 | 10 | 2504 | |||
XM_017016656.1 | XP_016872145.1 | 11 | 2566 | |||
XM_017016659.1 | XP_016872148.1 | 10 | 2473 | |||
GenBank mRNA | AK026182.1 | - | 6 | 1510 | ||
BC057787.1 | - | 6 | 1682 |