Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320210.1 | NP_001307139.1 | 4 | 569 | ||
NM_001321238.1 | NP_001308167.1 | 2 | 433 | |||
NM_001321239.1 | NP_001308168.1 | 2 | 1283 | |||
NM_024818.4 | NP_079094.1 | 4 | 1419 | |||
NM_198329.3 | NP_938143.1 | 4 | 442 | |||
GenBank mRNA | AB154406.1 | - | 4 | 378 | ||
AK026904.1 | - | 4 | 612 | |||
AK027563.1 | - | 3 | 211 | |||
AK056238.1 | - | 4 | 1420 | |||
AK129616.1 | - | 5 | 478 | |||
AL136757.1 | - | 4 | 432 | |||
AL832669.1 | - | 1 | 1638 | |||
AY253672.1 | - | 4 | 571 | |||
BC009737.1 | - | 4 | 548 | 3879061 | ||
BC048015.1 | - | 4 | 526 | |||
BI562266.1 | - | 2 | 439 | |||
DA042045.1 | - | 2 | 386 | |||
DA785394.1 | - | 4 | 532 | |||
DB457433.1 | - | 4 | 443 |