Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATRX SIRNA Assays ›
Gene Symbol:
ATRX
Gene Name:
ATRX, chromatin remodeler
Gene Aliases:
ATR2, JMS, MRX52, MRXHF1, RAD54, RAD54L, SFM1, SHS, XH2, XNP, ZNF-HX
Chromosome Location:
Chr.X: 77504878 - 77786269 on Build GRCh38
UniGene ID:
Hs.533526
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
3171
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATRX
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000489.4
NP_000480.3
9
1029
NM_138270.3
NP_612114.2
8
915
XM_005262153.4
XP_005262210.2
9
1026
XM_005262154.4
XP_005262211.2
9
1029
XM_005262155.4
XP_005262212.2
9
1130
XM_005262156.4
XP_005262213.2
8
882
XM_005262157.4
XP_005262214.2
8
912
XM_006724666.3
XP_006724729.1
8
912
XM_006724667.3
XP_006724730.1
7
768
XM_006724668.2
XP_006724731.1
9
1029
XM_017029601.1
XP_016885090.1
9
1026
XM_017029602.1
XP_016885091.1
9
1110
XM_017029603.1
XP_016885092.1
8
879
XM_017029604.1
XP_016885093.1
8
915
XM_017029605.1
XP_016885094.1
9
1130
XM_017029606.1
XP_016885095.1
8
1016
XM_017029607.1
XP_016885096.1
8
1013
XM_017029608.1
XP_016885097.1
7
765
XM_017029609.1
XP_016885098.1
8
1016
XM_017029610.1
XP_016885099.1
8
1013
XM_017029611.1
XP_016885100.1
7
768
GenBank mRNA
AB102641.1
-
9
761
AB208928.1
-
8
843
AK307791.1
-
8
855
AK308419.1
-
9
968
U72936.2
-
10
1098
U72937.2
-
9
976
U72938.2
-
8
862
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
gene-specific transcriptional regulator
Gene Ontology Categories:
Function(s)
Process(es)
DNA repair
DNA methylation
DNA recombination
nucleosome assembly
DNA replication-independent nucleosome assembly
chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
spermatogenesis
positive regulation of nuclear cell cycle DNA replication
covalent chromatin modification
DNA damage response, signal transduction by p53 class mediator
forebrain development
replication fork processing
positive regulation of telomere maintenance
DNA duplex unwinding
post-embryonic forelimb morphogenesis
multicellular organism growth
positive regulation of transcription from RNA polymerase II promoter
Sertoli cell development
protein localization to chromosome, telomeric region
seminiferous tubule development
cellular response to hydroxyurea
regulation of histone H3-K9 trimethylation
negative regulation of telomeric RNA transcription from RNA pol II promoter
positive regulation of telomeric RNA transcription from RNA pol II promoter
negative regulation of maintenance of mitotic sister chromatid cohesion, telomeric
DNA binding
DNA helicase activity
chromatin binding
helicase activity
protein binding
ATP binding
DNA translocase activity
methylated histone binding
histone binding
metal ion binding
chromo shadow domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0