Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIPAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321635.1 | NP_001308564.1 | 8, 9 | 1144 | ||
NM_001321636.1 | NP_001308565.1 | 8, 9 | 1144 | |||
NM_024759.2 | NP_079035.1 | 8, 9 | 1144 | |||
NR_135745.1 | 9, 10 | 1308 | ||||
NR_135746.1 | 10, 11 | 1430 | ||||
NR_135747.1 | 9, 10 | 1368 | ||||
GenBank mRNA | AK024017.1 | - | 8, 9 | 1144 | ||
AK302414.1 | - | 7, 8 | 907 | |||
BC132687.1 | - | 8, 9 | 1087 | 40146709 | ||
BC144055.1 | - | 8, 9 | 1087 | 9052575 |