Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160364.2 | NP_001153836.1 | 8 | 1285 | ||
NM_001300732.1 | NP_001287661.1 | 8 | 1187 | |||
NM_001324263.1 | NP_001311192.1 | 8 | 1174 | |||
NM_001324265.1 | NP_001311194.1 | 8 | 1187 | |||
NM_001324268.1 | NP_001311197.1 | 7 | 1039 | |||
NM_024847.4 | NP_079123.3 | 8 | 1187 | |||
NR_136733.1 | 8 | 1174 | ||||
GenBank mRNA | AK024893.1 | - | 4 | 396 | ||
AK094664.1 | - | 8 | 1187 | |||
AK293874.1 | - | 8 | 1081 | |||
AY236498.1 | - | 8 | 1121 | |||
AY263165.1 | - | 8 | 1116 | |||
BC036205.1 | - | 8 | 1116 | 5261157 | ||
BC047719.1 | - | 8 | 1118 | 5743899 | ||
CR933669.1 | - | 8 | 1285 |