Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2H4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001517.4 | NP_001508.1 | 3, 4 | 458 | ||
GenBank mRNA | AK222607.1 | - | 3, 4 | 362 | ||
AK298058.1 | - | 3, 4 | 506 | |||
AK300239.1 | - | 2, 3 | 311 | |||
AK309869.1 | - | 3, 4 | 472 | |||
BC004935.1 | - | 3, 4 | 430 | 3606718 | ||
BC016302.2 | - | 3, 4 | 430 | 3830902 | ||
BI460355.1 | - | 3, 4 | 479 | |||
BT007321.1 | - | 2, 3 | 251 | |||
Y07595.1 | - | 3, 4 | 378 |