Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002292.3 | NP_001002292.3 | 4 | 768 | ||
NM_001193334.1 | NP_001180263.1 | 3 | 501 | |||
NM_024911.6 | NP_079187.3 | 4 | 829 | |||
XM_011542191.2 | XP_011540493.1 | 4 | 752 | |||
XM_011542192.2 | XP_011540494.1 | 4 | 762 | |||
XM_017002390.1 | XP_016857879.1 | 4 | 596 | |||
GenBank mRNA | AK074583.1 | - | 4 | 784 | ||
AK074984.1 | - | 4 | 751 | |||
AK309779.1 | - | 5 | 708 | |||
AY359035.1 | - | 4 | 811 | |||
BC007211.1 | - | 3 | 367 | |||
BC110826.1 | - | 4 | 730 | 6183768 | ||
BC137109.1 | - | 4 | 626 | 9021106 | ||
BC137113.1 | - | 4 | 626 | 9021110 | ||
BX538320.1 | - | 4 | 768 | |||
BX648748.1 | - | 3 | 499 | |||
DQ323735.1 | - | 4 | 798 |