Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGIX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099681.2 | NP_001093151.2 | 5 | 1043 | ||
NM_001099682.2 | NP_001093152.2 | 5 | 1028 | |||
NM_024859.3 | NP_079135.3 | 6 | 1271 | |||
XM_005278062.4 | XP_005278119.1 | 6 | 1310 | |||
XM_005278063.4 | XP_005278120.1 | 6 | 1294 | |||
XM_005278064.4 | XP_005278121.1 | 4 | 1204 | |||
XM_005278065.3 | XP_005278122.1 | 4 | 1292 | |||
XM_006724560.3 | XP_006724623.1 | 6 | 1340 | |||
XM_011543985.2 | XP_011542287.1 | 5 | 1425 | |||
XM_011543986.2 | XP_011542288.1 | 4 | 1179 | |||
XM_011543987.2 | XP_011542289.1 | 4 | 1200 | |||
GenBank mRNA | AK025340.1 | - | 4 | 1126 | ||
AK096058.1 | - | 4 | 2351 | |||
BC067800.1 | - | 5 | 1044 |