Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM192A SIRNA Assays ›
Gene Symbol:
FAM192A
Gene Name:
family with sequence similarity 192 member A
Gene Aliases:
C16orf94, CDA018, CDA10, NIP30
Chromosome Location:
Chr.16: 57152466 - 57186242 on Build GRCh38
UniGene ID:
Hs.396740
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
32078
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM192A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_024946.2
NP_079222.1
3, 4
477
XM_005256156.4
XP_005256213.1
3, 4
445
XM_005256158.3
XP_005256215.1
3, 4
1532
XM_005256160.2
XP_005256217.1
3, 4
449
XM_005256163.2
XP_005256220.1
4, 5
586
XM_005256164.3
XP_005256221.1
4, 5
531
XM_005256165.2
XP_005256222.1
3, 4
506
XM_006721275.3
XP_006721338.2
3, 4
1215
XM_011523343.2
XP_011521645.1
3, 4
323
XM_017023681.1
XP_016879170.1
3, 4
541
XM_017023682.1
XP_016879171.1
4, 5
1650
XM_017023683.1
XP_016879172.1
4, 5
1601
XM_017023684.1
XP_016879173.1
4, 5
816
XM_017023685.1
XP_016879174.1
4, 5
1565
XM_017023686.1
XP_016879175.1
5, 6
1635
XM_017023687.1
XP_016879176.1
5, 6
1610
XM_017023688.1
XP_016879177.1
5, 6
1553
XM_017023689.1
XP_016879178.1
5, 6
934
XM_017023690.1
XP_016879179.1
5, 6
885
XM_017023691.1
XP_016879180.1
5, 6
938
XM_017023692.1
XP_016879181.1
4, 5
722
XM_017023693.1
XP_016879182.1
4, 5
625
XM_017023694.1
XP_016879183.1
5, 6
728
XM_017023695.1
XP_016879184.1
5, 6
840
XM_017023696.1
XP_016879185.1
4, 5
1223
XM_017023697.1
XP_016879186.1
4, 5
608
XM_017023698.1
XP_016879187.1
4, 5
1331
XM_017023699.1
XP_016879188.1
4, 5
859
XM_017023700.1
XP_016879189.1
4, 5
328
XM_017023701.1
XP_016879190.1
4, 5
794
XM_017023702.1
XP_016879191.1
4, 5
514
XM_017023703.1
XP_016879192.1
3, 4
254
XM_017023704.1
XP_016879193.1
3, 4
252
GenBank mRNA
AF212249.1
-
3, 4
430
AF271785.1
-
3, 4
430
AF356585.1
-
2, 3
228
AK023291.1
-
3, 4
377
AK025452.1
-
3, 4
452
AL834421.1
-
3, 4
289
BC020536.1
-
3, 4
445
3849620
BC063409.1
-
3, 4
385
BC071952.1
-
2, 3
246
6305656
BI545617.1
-
3, 4
482
DB101263.1
-
3, 4
452
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-cq5mm:80/10.239.166.46:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0