Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MCF2L SIRNA Assays ›
Gene Symbol:
MCF2L
Gene Name:
MCF.2 cell line derived transforming sequence like
Gene Aliases:
ARHGEF14, DBS, OST
Chromosome Location:
Chr.13: 112894378 - 113099739 on Build GRCh38
UniGene ID:
Hs.170422
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
32285
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MCF2L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001112732.2
NP_001106203.2
17
2111
NM_001320815.1
NP_001307744.1
17
2143
NM_001320816.1
NP_001307745.1
17
2320
NM_001320817.1
NP_001307746.1
17
2143
NM_024979.4
NP_079255.4
17
2143
XM_011537482.1
XP_011535784.1
18
2100
XM_011537483.2
XP_011535785.1
18
2220
XM_011537484.1
XP_011535786.1
17
2547
XM_011537485.1
XP_011535787.1
17
2219
XM_011537486.2
XP_011535788.2
17
2235
XM_011537487.1
XP_011535789.1
17
2140
XM_011537488.1
XP_011535790.1
17
1979
XM_011537490.1
XP_011535792.1
17
2513
XM_011537491.1
XP_011535793.1
15
1928
XM_017020493.1
XP_016875982.1
17
2235
XM_017020494.1
XP_016875983.1
17
2235
XM_017020495.1
XP_016875984.1
17
2235
XM_017020496.1
XP_016875985.1
15
1928
XM_017020497.1
XP_016875986.1
17
2513
XM_017020498.1
XP_016875987.1
15
1928
XM_017020499.1
XP_016875988.1
17
2235
GenBank mRNA
AB002360.1
-
17
2088
AB116074.1
-
17
1999
AB116075.1
-
12
1475
AK094768.1
-
15
1908
AK295047.1
-
17
2140
AK295670.1
-
17
1953
AK315969.1
-
17
2111
BC011853.2
-
17
2103
4138009
BC020208.1
-
17
2103
4121441
Back To Top
More Information
Panther Classification:
Molecular Function -
guanyl-nucleotide exchange factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of Rho protein signal transduction
intracellular signal transduction
positive regulation of apoptotic process
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
phosphatidylinositol binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0