Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270367.1 | NP_001257296.1 | 4 | 519 | ||
NM_001270368.1 | NP_001257297.1 | 4 | 532 | |||
NM_001270369.1 | NP_001257298.1 | 5 | 571 | |||
NM_001270370.1 | NP_001257299.1 | 6 | 578 | |||
NM_001270371.1 | NP_001257300.1 | 5 | 593 | |||
NM_001270372.1 | NP_001257301.1 | 4 | 511 | |||
NM_001270373.1 | NP_001257302.1 | 5 | 550 | |||
NM_001270374.1 | NP_001257303.1 | 5 | 900 | |||
NM_025125.3 | NP_079401.2 | 4 | 498 | |||
NR_072984.1 | 5 | 515 | ||||
NR_072985.1 | 5 | 522 | ||||
NR_072986.1 | 4 | 420 | ||||
NR_072987.1 | 5 | 472 | ||||
XM_006717978.2 | XP_006718041.1 | 5 | 850 | |||
GenBank mRNA | AK023325.1 | - | 4 | 474 | ||
AK222536.1 | - | 4 | 479 | |||
AK300490.1 | - | 4 | 529 | |||
AK309425.1 | - | 5 | 797 | |||
AK309980.1 | - | 4 | 506 | |||
AK310160.1 | - | 5 | 480 | |||
AK311563.1 | - | 5 | 488 | |||
AL832397.1 | - | 4 | 395 | |||
AL832483.1 | - | 4 | 506 | |||
BC022252.1 | - | 4 | 474 | 4668550 | ||
BP204746.1 | - | 6 | 547 | |||
BP293793.1 | - | 5 | 525 | |||
BX537803.1 | - | 5 | 480 | |||
CX756014.1 | - | 6 | 534 | |||
DA573956.1 | - | 5 | 531 | |||
DA930892.1 | - | 4 | 487 |