Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RUFY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040451.2 | NP_001035541.1 | 4 | 754 | ||
NM_001040452.2 | NP_001035542.1 | 5 | 565 | |||
NM_025158.4 | NP_079434.3 | 5 | 740 | |||
XM_005265993.4 | XP_005266050.1 | 2 | 111 | |||
XM_006714921.3 | XP_006714984.2 | 5 | 750 | |||
XM_017009891.1 | XP_016865380.1 | 5 | 565 | |||
XM_017009892.1 | XP_016865381.1 | 2 | 129 | |||
XM_017009893.1 | XP_016865382.1 | 2 | 129 | |||
XM_017009894.1 | XP_016865383.1 | 5 | 473 | |||
XM_017009895.1 | XP_016865384.1 | 4 | 545 | |||
GenBank mRNA | AB209507.1 | - | 5 | 465 | ||
AF312367.1 | - | 5 | 731 | |||
AF361055.2 | - | 5 | 713 | |||
AK075021.1 | - | 4 | 653 | |||
AK291926.1 | - | 4 | 754 | |||
BC032571.1 | - | 5 | 484 | 5243223 | ||
DA925727.1 | - | 5 | 526 |