Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: WNT10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025216.2 | NP_079492.2 | 1, 2 | 584 | ||
XM_011511930.1 | XP_011510232.1 | 1, 2 | 237 | |||
GenBank mRNA | AB059569.1 | - | 1, 2 | 126 | ||
AK024363.1 | - | 1, 2 | 584 | |||
AY009400.1 | - | 1, 2 | 317 | |||
BC034352.1 | - | 1, 2 | 243 | |||
BC040026.1 | - | 1, 2 | 554 | |||
BC052234.1 | - | 1, 2 | 595 | 5442628 |